VWR Life Science Cyclo-Prep™ PCR Clean Up Kit
Supplier: Avantor
VWR® Cyclo-Prep™
K600-50RXN
97063-566EA 178.36 USD
97063-566
VWR Life Science Cyclo-Prep™ PCR Clean Up Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The VWR® Cyclo-Prep™ PCR Clean Up Kit is a spin column-based purification kit for the rapid isolation of PCR fragments following amplification. After clean-up, ultra-pure isolated DNA may be used for restriction enzyme digestion, labeling, hybridization, ligation, DNA cloning and fluorescent sequencing protocols. The Cyclo-Prep™ PCR Clean Up Kit contains all necessary reagents for DNA purification with no hazardous phenol or chloroform, making it both safe and effective. Add 64 mL of Ethanol to Wash Buffer before use.
  • Rapid isolation of PCR fragments
  • Fast spin column format
  • Pure product ideal for cloning & sequencing
Ordering information: Kit includes Binding Buffer 55 mL, Wash Buffer 16 mL, Elution Buffer 10 mL, 50 Spin Columns, and 50 Collection Tubes. Contains sufficient reagents for 50 isolations.