Phenol saturated with buffer, Biotechnology Grade 1 Phase

Danger

Synonyms: Carbolic acidHydroxybenzenePhenylalcohol

K168-400ML
97064-822EA 158.91 USD
97064-822
Phenol saturated with buffer, Biotechnology Grade 1 Phase
Phenol

Phenol S/P mixtures are ready to use, single-phase phenol solutions that do not have to be equilibrated with a buffering solution prior to use. These phenol products for nucleic acid purification allow easier sample preparation and reduce the hazards associated with preparing traditional phenol solutions in the laboratory.


  • High quality & nuclease-free
  • Ready-to-use single-phase phenols
  • No equilibration required prior to use

Formula: C₆H₅OH
MW: 94.11 g/mol
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 2821
ADR: 6.1,II

Order Now

Specification Test Results

Conductivity(5%, Water) @25°C 180 uS
pH(5%, Water) @25°C 7.5 - 7.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR