Milk, non fat (skimmed milk), powder, Proteomics Grade
Supplier: VWR
M203-10G-10PK
97063-958EA
99.95
USD
97063-958
Milk, non fat (skimmed milk), powder, Proteomics Grade
Non-fat milk powder (skim milk)
10 g per pack
Storage Temperature:
Ambient |
CAS Number:
999999-99-4 EINECS: 310-127-6 |
Specification Test Results
Moisture (LOD) |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...