Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3'-Diaminobenzidine (DAB) substrate, tablets for immunohistochemistry

Supplier: VWR
Danger
Synonyms: DAB3,3',4,4'-BiphenyltetraminBiphenyl-3,3',4,4'-tetrayltetraamine

E733-100TABS E733-50TABS E733-50TABS E733-100TABS
97062-970EA 158.19 USD InStock
97062-970 97062-972 CA97062-972 CA97062-970
3,3'-Diaminobenzidine (DAB) substrate, tablets for immunohistochemistry
3,3'-Diaminobenzidine
Commonly used for detection of horseradish peroxidase in membrane-bound and In Situ applications

Insoluble end product is resistant to alcohol and fading, making it ideal for immunohistochemistry. Each tablet contains 5 mg of DAB.


Formula: C₁₂H₁₄N₄
MW: 214,27 g/mol
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6
Order Now
Specification Test Results
Immunoperoxidase Assay PASS / Fail

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty