Amphotericin B for tissue culture

Warning

E437-1G
97062-758EA 403.94 USD
97062-758
Amphotericin B for tissue culture
Amphotericin B

An antimycotic agent that acts by altering membrane permeability. Susceptible organisms: yeast, molds. Working concentration: 2.5 μg/ml

Formula: C₄₇H₇₃NO₁₇
MW: 924.08 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00877763
CAS Number: 1397-89-3
EINECS: 215-742-2

Order Now

Specification Test Results

Activity mcg/mg is lot dependent

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR