D-(+)-Galactose ≥99.0%, high purity

Synonyms: (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal(2R,3S,4S,5R)-Pentahydroxyhexanal

0637-100G 0637-500G
97062-076CS 907.1 USD
97062-076 97062-080
D-(+)-Galactose ≥99.0%, high purity
D(+)-Galactose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527 °C (1013 hPa)
Melting Pt: 163…169 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://us.vwr.com/stibo/web/std.lang.all/21/21/2302121.jpg

Order Now

Specification Test Results

Purity = 99
Specific Rotation (2%, Water 5 Hrs.) 79° - 81°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR