L-Arginine hydrochloride USP

Synonyms: (S)-(+)-Arginine hydrochlorideL-ArgL-Arginine, hydrochloride(S)-2-Amino-5-guanidinopentanoic acid hydrochlorideL(+)-Arginine hydrochlorideL(+)-Arginine monohydrochloride(S)-(+)-Arginine HClH-L-Arg-OH.HClH-Arg-OH.HCl

0877-500G 0877-100G
97062-468CS 1847.71 USD
97062-468 97062-466
L-Arginine hydrochloride USP
L-Arginine hydrochloride
Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 218 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR