L(+)-Valine USP

Synonyms: (S)-(+)-ValineValineH-Val-OH

1B1102-100G
97062-562CS 1077.85 USD
97062-562
L(+)-Valine USP
L(+)-Valine

Non-animal origin. White solid.

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR