Octyl-β-D-thioglucopyranoside
Supplier: VWR
Synonyms:
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol
, N-Octyl-1-thio-Beta-D-glucopyranoside
, OTG
, 1-S-Octyl-β-D-thioglucopyranoside
, Octyl-thioglucoside
J575-1G
97063-228EA
113.07
USD
97063-228
Octyl-β-D-thioglucopyranoside
Octyl β-D-thioglucopyranoside
Non-ionic - containing no charge (polyoxyethylenes or alkyl glucosides). Non-ionic detergents are suited for solubilizing membrane proteins without altering biological activity. These non-denaturing, gentler detergents are ideal for studying the conformat
Formula:
C₁₄H₂₈O₅S MW: 308.44 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00012189 CAS Number: 85618-21-9 |
Specification Test Results
Purity |
|
Solubility (5%, Water) | Pass |
Specific Rotation (1%, MeOH) | (- 55°) - (-49°) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...