TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid) free acid ≥99%, high purity

Synonyms: N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulfonic acidN-Tris(hydroxymethyl)methyl-3-aminopropanesulfonic acid

J562-100G
97064-208CS 704.76 USD
97064-208
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid) free acid ≥99%, high purity
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, and not interfere with biological processes.

Formula: C₇H₁₇NO₆S
MW: 243.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007538
CAS Number: 29915-38-6
EINECS: 249-954-1

Order Now

Specification Test Results

DNase none detected
Heavy Metals = 0.0005 %
Appearance White powder
pH (5%, Water) @25°C 3.5 - 6.5
Protease none detected
Purity = 99 %
RNase none detected
Solubility (5%, Water) Pass
Water (KF) = 1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR