Magnesium sulfate, anhydrous USP

Synonyms: Magnesium(II) sulfateBitter saltSulfuric acid magnesium salt (1:1)Epsom salt

E797-2.5KG E797-500G
97062-996CS 1022.65 USD
97062-996 97062-998
Magnesium sulfate, anhydrous USP
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://us.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg

Order Now

Specification Test Results

Chloride 0.014%
Heavy Metals 0.001 %
Iron 0.002 %
Loss on Drying 2 %
pH (5%, Water) @25°C 5 - 9.2
Purity (after ignition) 99 % - 100.5 %
Selenium 0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR