Ampicillin sodium salt for tissue culture

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

E477-20ML
97062-796EA 52.32 USD
97062-796
Ampicillin sodium salt for tissue culture
Ampicillin sodium salt

  • Formula: C16H18N3NaO4S
  • pH (1%, Water) @25C: 8.0-10.0
  • Potency (Anhydrous) (mcg/mg): 845-988


These antibiotic and antimycotic mixtures have been aseptically prepared from the highest quality materials and are gamma-irradiated to ensure sterility. The dry blends, or lyophilized powders, are packaged in amber serum vials with multiple injection stoppers for syringe mediated liquid handling. Simply reconstitute the powder in the recommended volume of sterile tissue culture grade water. Each reagent has been tested for compatibility with standard tissue culture systems and remains stable for greater than one year when stored as recommended. All powders and solutions should be stored at 4C unless otherwise noted.


Aseptically prepared and gamma irradiated to ensure sterility. Functionally tested for compatibility with standard tissue culture systems. When reconstituted in 20 mL sterile water, contains 10 mg/mL Ampicillin.

Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR