Nonidet® P 40 Substitute (NP-40), Reagent Grade

Warning

E109-100ML E109-500ML
97064-730EA 49.99 USD
97064-730 97064-732
Nonidet® P 40 Substitute (NP-40), Reagent Grade
Nonidet® P 40 Substitute (NP-40)

This non-ionic detergent is suitable for solubilizing membrane proteins without altering biological activity.

Boiling Pt: ∼290 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: >150 °C
EINECS: 500-024-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Density (20°C) 1.05 g/ml - 1.07 g/ml
Refractive Index 1.48 - 1.52
Solubility (5%, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR