Cycloheximide, crystalline, Ultra Pure Grade

Supplier: VWR
Danger

Synonyms: 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,Naramycin AActidione

94271-25G 94271-1G 94271-100MG 94271-5G
97064-728EA 198.27 USD
97064-728 97064-722 97064-724 97064-726
Cycloheximide, crystalline, Ultra Pure Grade
Cycloheximide
Formula: C₁₅H₂₃NO₄
MW: 281.35 g/mol
Boiling Pt: 358 °C (1013 hPa)
Melting Pt: 119.5…121 °C
MDL Number: MFCD00082346
CAS Number: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1,I
Merck Index: 12,02797
https://us.vwr.com/stibo/web/std.lang.all/89/03/2298903.jpg

Order Now

Specification Test Results

Assay >97.0%
Specific optical rotation -32 ...-25 °
Loss on drying <0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR