Potassium acetate, anhydrous USP

Synonyms: Diuretic salt4-Chloro-N-furfuryl-5-sulfamoylanthranlic acidAcetic acid potassium salt

0942-100G
97062-532CS 590.38 USD
97062-532
Potassium acetate, anhydrous USP
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304 °C
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 12,07764
https://us.vwr.com/stibo/web/std.lang.all/05/72/16640572.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR