Sodium nitrate, Reagent Grade

Warning

0925-1KG
97064-996CS 288.08 USD
97064-996
Sodium nitrate, Reagent Grade
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 12,08792

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.001%
Heavy Metals = 0.0005%
Insolubles = 0.005%
Iodate = 0.0005%
Iron = 0.0003%
Magnesium = 0.002%
Nitrite = 0.001%
pH (5%, Water) @25°C 5.5 - 8.3
Phosphate = 0.0005%
Purity = 99%
Sulfate = 0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR