Sudan black B

Supplier: VWR

Synonyms: Ceres black BNSolvent Black 3CI 26150Fat Black HB

0593-50G
97061-984EA 100.47 USD
97061-984
Sudan black B
Sudan black B

A lipid stain used for demonstrating triglycerides in frozen sections. Also a histochemical stain for chromosomes, Golgi apparatus, and leucocyte granules.

Formula: C₂₉H₂₄N₆
MW: 456.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006919
CAS Number: 4197-25-5
EINECS: 224-087-1

Order Now

Specification Test Results

Lambda Max 596 - 605 nm
Melting Point 118 - 126 °C
P-15/P+15 0.98 - 1.02

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR