Methyl-4-hydroxybenzoate NF

Warning

Synonyms: Methyl ParahydroxybenzoateMethyl 4-hydroxybenzoate4-Hydroxybenzoic acid methyl esterMethylparaben

0572-500G
97061-946EA 76.25 USD
97061-946
Methyl-4-hydroxybenzoate NF
Methyl 4-hydroxybenzoate

Fine white crystalline powder

Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 270…280 °C (1013 hPa)
Melting Pt: 125…128 °C
Density: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 12,06182
https://us.vwr.com/stibo/web/std.lang.all/17/24/2301724.jpg

Order Now

Specification Test Results

Acidity Test Pass
Color of Solution Pass
Purity 98.0 - 102.0 %
Related Substances Test Pass
Residue on Ignition = 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR