Brilliant green
Supplier: VWR
|
Warning
|
Synonyms:
CI 42040
, Basic Green 1
, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulfate
, Brilliant Green hydrogen sulfate
, Brilliant Green hydrogen sulphate
, Diamond Green
, Astradiamant green GX
, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate
, Solid Green JO
, Ethyl Green
, Malachite Green G
, Emerald Green
A stain for differentation of E. coli from S. typhosa. Also used in bacteriological media.Color index: 42040
Formula:
C₂₇H₃₄N₂O₄S MW: 482.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011880 CAS Number: 633-03-4 EINECS: 211-190-1 |
Specification Test Results
Absorbance @ lambda Max |
|
Dye Content |
|
Lambda max (.003 mg/ml 50% Ethanol) | 628 - 632 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...