Tetracycline hydrochloride USP

Warning

0422-25G 0422-100G VWRV0422-EU-25G
97061-640EA 45.09 USD
97061-640 97061-638 75856-696
Tetracycline hydrochloride USP
Tetracycline hydrochloride

Recommended working concentration: 15 µg/ml


Inhibits protein synthesis in gram-positive and gram-negative bacteria.

Formula: C₂₂H₂₅ClN₂O₈
MW: 480.9 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00078142
CAS Number: 64-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR