Nystatin

Warning

Synonyms: MycostatinFungicidin

0418-25MU
97061-630EA 49.13 USD
97061-630
Nystatin
Nystatin

Nystatin is antimyctotic agent that alters membrane permeability in a broad spectrum of yeasts and molds.

Formula: C₄₇H₇₅NO₁₇
MW: 926.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD25541651
CAS Number: 1400-61-9
EINECS: 215-749-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR