Guanosine for biochemistry

Supplier: VWR
Danger

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

0193-100G
97061-190CS 2596.33 USD
97061-190
Guanosine for biochemistry
Guanosine
Formula: C₁₀H₁₃N₅O₅
MW: 283.24 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
EINECS: 204-227-8

Order Now

Specification Test Results

Em(252.5nm,0.01M PO4 buffer) 12500 - 15000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR