    Sort by:
  • Search Within Results
3-methylnonane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (TCM0282-005ML)
Supplier:  TCI America
Description:   CAS Number: 5911-04-6 MDL Number: MFCD00027308 Molecular Formula: C10H22 Molecular Weight: 142.29 Purity/Analysis Method: 98.0% (GC) Form: Cl...