    Sort by:
  • Search Within Results
2-Methyl-4-heptanone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (TCM0694-005ML)
Supplier:  TCI America
Description:   CAS Number: 626-33-5 MDL Number: MFCD00027143 Molecular Formula: C8H16O Molecular Weight: 128.22 Purity/Analysis Method: 99.0% (GC) Form: Cle...