    Sort by:
  • Search Within Results
3,3-Bis(bromomethyl)pentane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   3,3-Bis(bromomethyl)pentane. Grade:tech. 90, Melting Point C38-41*. Boiling Point C:96-97*/10mm. C7H14Br2. 67969-84-0. IRRITANT