You Searched For: 1,2-Propanedithiol
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: TCI America
Description: CAS Number: 814-67-5 MDL Number: MFCD00022073 Molecular Formula: C3H8S2 Molecular Weight: 108.22 Purity/Analysis Method: 95.0% (GC) Form: Cle...