    Sort by:
1-Pentanol
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"11"
  List View Easy View
Sort by:
Supplier:  Avantor Performance Materials
Description:   Amyl alcohol; pentyl alcohol. CAS RN 71-41-0. Formula Weight: 88.15. Density: 0.82 kg/L. AR*, 98.0% min. by GC. Boiling Range: 135-139[degree]C. Residue after Evaporation: 0.003% max. Actual signed Certificate of Lot Analysis provided on label. Postal/shipping restrictions. 4L.
SDS
Supplier:  MilliporeSigma
Description:   Cas Number 71-41-0 1L
SDS
Supplier:  MilliporeSigma
Description:   N-AMYL ALCOHOL (PENTAN-1- (1L)
SDS
Supplier:  TCI America
Description:   CAS Number: 71-41-0 MDL Number: MFCD00002977 Molecular Formula: C5H12O Molecular Weight: 88.15 Purity/Analysis Method: 99.0% (GC) Form: Clear...
SDS
Supplier:  Alfa Aesar
Description:   98+%. 500mL.
SDS
Supplier:  Alfa Aesar
Description:   ACS, 99+%,UN1105,Liquid,DANGER: FLAMMABLE, causes narcosis, irritation,4L,1L,FLAMMABLE LIQUID,
SDS
Supplier:  BeanTown Chemical
Description:   CAS: 71-41-0; EC No: 200-752-1; MDL No: MFCD00002977; RTECS: SB9800000 UN No: UN1105; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3(CH2...
SDS
Supplier:  BeanTown Chemical
Description:   CAS: 71-41-0; EC No: 200-752-1; MDL No: MFCD00002977; RTECS: SB9800000 UN No: UN1105; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3(CH2...
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 11
  1  2  Next