You Searched For: 1-Octanol


16  results were found

SearchResultCount:"16"

Sort Results

List View Easy View

Rate These Search Results

Supplier: MilliporeSigma
Description: 1-OCTANOL FOR SYNTHESIS (1L)
Supplier: MilliporeSigma
Description: Emplura*1-Octanol, Cas number: 111-87-5, Chemical Formula:CH3(CH2)7OH, Solvent for lab application, 1l.
Supplier: MilliporeSigma
Description: Capryl alcohol; caprylic alcohol; octyl alcohol; n-octyl alcohol. CAS RN 111-87-5. Formula Weight: 130.23. 98% min. by GC. 4L.
Supplier: Thermo Scientific Chemicals
Description: 1-OCTANOL 99% PURE 250ML

Supplier: Thermo Scientific Chemicals
Description: 1-OCTANOL ACS REAGENT 1L

Supplier: Thermo Scientific Chemicals
Description: 1-OCTANOL 99% ANHYDROUS ACROSEAL 1L

Supplier: Thermo Scientific Chemicals
Description: 99%. 2500mL.
Supplier: Thermo Scientific Chemicals
Description: 1-OCTANOL NATURAL 98% 111-87-5 10L
Supplier: TCI America
Description: CAS Number: 111-87-5 MDL Number: MFCD00002988 Molecular Formula: C8H18O Molecular Weight: 130.23 Purity/Analysis Method: 99.0% (GC) Form: Cle...
Catalog Number: (TCO0212-500ML)
Supplier: TCI America
Description: [for Determining Partition Coefficients] CAS Number: 111-87-5 MDL Number: MFCD00002988 Molecular Formula: C8H18O Molecular Weight: 130.23 Purity/...

Catalog Number: (700002-694)
Supplier: Spectrum Chemicals
Description: 1-Octanol, Reagent, ACS is a fatty alcohol that functions as a fragrance ingredient. It is a liquid and is insoluble in cold water. As an ACS grade re...


Catalog Number: (BJ472328-1L)
Supplier: Honeywell Research Chemicals
Description: ACS GENERAL USE SOLVENTS

Catalog Number: (BJ112615-2.5L)
Supplier: Honeywell Research Chemicals
Description: 1-Octanol, General Purpose Reagent, CAS Number: 111-87-5, Purity: >/= 99%, Synonyms: Alcohol C8, Capryl alcohol, Octyl alcohol, Performance Grade Solvent, Reagent Grade, Size: 2.5L

SDS


Supplier: BeanTown Chemical
Description: 1-Octanol, Purity: 99%, CAS no: 111-87-5, Molecular Formula: C8H18O, Synonyms: :Capryl alcohol, n-Octyl alcoholAlcohol C8, Size: 500ML

SDS

Catalog Number: (BJ293245-1L)
Supplier: Honeywell Research Chemicals
Description: 1-Octanol, Purity: >/=99%, Grade: Chromasolv, for HPLC, CAS Number: 111-87-5, MF: CH3(CH2)7OH, Molar mass: 130.23 g/mol, Synonyms: Alcohol C8, Capryl alcohol, Octyl alcohol, High Purity Solvent, Liquid Chromatography, 1L, Container Type: Glass bottle, Appearance: Colorless, Size: 1L

Supplier: Ambeed
Description: n-Octanol, Purity: 98%, CAS number: 111-87-5, Appearance: Form: liquid Colour: colourless, Storage: Sealed in dry, Room Temperature, Size: 100G

New Product

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 16
no targeter for Bottom