    Sort by:
  • Search Within Results
butyl-isocyanate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (AAL05664-14)
Supplier:  Alfa Aesar
Description:   1-Butyl isocyanate, Purity: 97%, Cas No: 111-36-4, Molecular Formula: C5H9NO, Color: colourless, Form: Liquid, 25g