You Searched For: Tetraiodomethane
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (TCC0936-010G)
Supplier: TCI America
Description: CAS Number: 507-25-5 MDL Number: MFCD00001067 Molecular Formula: CI4 Molecular Weight: 519.63 Purity/Analysis Method: 97.0% (T) Form: Crystal...