You Searched For: Propionic acid


14  results were found

SearchResultCount:"14"

Sort Results

List View Easy View

Rate These Search Results

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Propionic Acid, BAKER
Catalog Number: (JTU340-09)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Propionic Acid, BAKER

Supplier: MilliporeSigma
Description: Propionic acid for synthesis, CAS number 79-09-4, Chemical formula CH3CH2COOH, Synonyms Methyl acetic acid 100ml
Catalog Number: (EM8.00605.9025)
Supplier: MilliporeSigma
Description: Propionic acid for synthesis, CAS number 79-09-4, Chemical formula CH3CH2COOH, Synonyms Methyl acetic acid 25l

Supplier: TCI America
Description: CAS Number: 79-09-4 MDL Number: MFCD00002756 Molecular Formula: C3H6O2 Molecular Weight: 74.08 Purity/Analysis Method: 99.0% (GC,T) Form: Cle...
Supplier: Thermo Scientific Chemicals
Description: Grade: 99+, Melting Point C-21*. Boiling Point C: 140-141*. C3H6O2. 79-09-4. CORROSIVE
Catalog Number: (IC15195680)
Supplier: MP Biomedicals
Description: PROPIONIC ACID (FRE 100.00ML

SDS


Supplier: Spectrum Chemicals
Description: Propionic Acid, FCC is used as a preservative and flavoring agent in packaged foods such as baked goods and cheese. The FCC grade meets the requiremen...
Catalog Number: (BJ49916-50ML)
Supplier: Honeywell Research Chemicals
Description: Propionic acid, Purity: >/=99.5% (GC), Grade: Analytical, Cas number: 79-09-4, Molecular Formula: CH3CH2COOH, Molar mass: 74.08 g/mol, Synonyms: Acid C3; Propanoic acid; Propanyl acid, Eluent additive for LC-MS, Size: 50ML

Catalog Number: (95031-450)
Supplier: Spectrum Chemicals
Description: Propionic Acid, NF is used as a preservative in pharmaceutical formulations. Spectrum Chemical manufactured NF grade products are manufactured, packag...

SDS Small Business Enterprise


Supplier: Thermo Scientific Chemicals
Description: PROPIONIC ACID 99+% EXTRA PURE 2500ML

Supplier: Thermo Scientific Chemicals
Description: PROPIONIC ACID ACS REAGENT 100ML

Supplier: Thermo Scientific Chemicals
Description: PROPIONIC ACID 1L

Supplier: BeanTown Chemical
Description: Propionic acid, Purity: 99%, CAS Number: 79-09-4, Molecular Formula: CH3CH2COOH, Synonyms: Propanoic acid, Propanyl acid, Size: 2.5kg

SDS

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 14 of 14
no targeter for Bottom