You Searched For: Acetic acid


102  results were found

SearchResultCount:"102"

Sort Results

List View Easy View

Rate These Search Results

Supplier: BDH
Description: BDH Acetic Acid, Glacial, ACS Grade. Meets Reagent Specifications for testing USP/NF monographs Size: 2.5L.
Catalog Number: (BDH7236-1)
Supplier: VWR International
Description: Standardized at 25 °C against chemicals whose certification is traceable to NIST. Made with high purity acid and deionized water. Suitable as a titran...

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, U.S.P., Multi-Compendial
Supplier: VWR
Description: GLACIAL ACETIC ACID ACS GRADE 2.5L
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, U.S.P. - F.C.C.
Supplier: BDH
Description: High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.
Catalog Number: (JT6903-5)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, ULTREX® II Ultrapure Reagent

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: For trace metals analysis, ICP, environmental, pharmaceutical and low metals.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, CMOS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, U.S.P., F.C.C., A.C.S.
Catalog Number: (MK312146)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, U.S.P., F.C.C., A.C.S.

Catalog Number: (MK248844)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, 36%, N.F.

Catalog Number: (JT9513-7)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Acetic Acid, Glacial, CMOS

Catalog Number: (CA71006-424)
Supplier: Anachemia
Description: ACETIC ACID GLACIAL ACS 2.5L

Catalog Number: (JT9524-51)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: For trace metal analysis. Certificate of lot analysis provided.

Catalog Number: (JT9515-3)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromato...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 102
no targeter for Bottom