You Searched For: Butyl acetate


18  results were found

SearchResultCount:"18"

Sort Results

List View Easy View

Rate These Search Results

Supplier: MilliporeSigma
Description: Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Supplier: MilliporeSigma
Description: Emsure* n-Butyl acetate for analysis, Cas number: 123-86-4, Chemical Formula:CH3COO(CH2)3CH3, Solvent, 4ml.
Supplier: MilliporeSigma
Description: Emplura* n-Butyl acetate, Cas number: 123-86-4, Chemical Formula:CH3COO(CH2)3CH3, Solvent for lab application25l.
Supplier: Thermo Scientific Chemicals
Description: n-Butyl acetate, Purity: 99.5+%Grade: ACS, Cas Number: 123-86-4, Molecular Formula: C6H12O2, Molecular Weight: 116.16, Density: 0.881, Melting point: -78 deg C, Boiling point: 124-126 deg C, Flash point: 25 deg C, Appearance: Liquid, Size: 4l
Supplier: Thermo Scientific Chemicals
Description: Semiconductor Grade, 99% min,UN1123,Liquid,DANGER: FLAMMABLE, irritates skin and eyes,4L,1L,FLAMMABLE LIQUID,
Catalog Number: (TCA0228-500ML)
Supplier: TCI America
Description: [for Spectrophotometry] CAS Number: 123-86-4 MDL Number: MFCD00009445 Molecular Formula: C6H12O2 Molecular Weight: 116.16 Purity/Analysis Method: ...

Supplier: TCI America
Description: CAS Number: 123-86-4 MDL Number: MFCD00009445 Molecular Formula: C6H12O2 Molecular Weight: 116.16 Purity/Analysis Method: 99.0% (GC) Form: Cl...
Supplier: Thermo Scientific Chemicals
Description: 99% 2.5L
Supplier: Spectrum Chemicals
Description: CAS: 123-86-4. Size: 20 Liter.
Supplier: Honeywell Research Chemicals
Description: ACS GENERAL USE SOLVENTS

Supplier: BeanTown Chemical
Description: N-Butyl acetate, Purity: 99%, CAS Number: 123-86-4, Molecular Formula: C6H12O2, Synonyms: Butyl acetate, Acetic acid n-butyl ester, Size: 500ML

SDS

Supplier: Thermo Scientific Chemicals
Description: HPLC Grade, 99.5+%,UN1123,Liquid,DANGER: FLAMMABLE, irritates skin and eyes,1L,4L,FLAMMABLE LIQUID,
Catalog Number: (TS10775-0010)
Supplier: Thermo Scientific Chemicals
Description: n-Butyl acetate, 99+%, extra pure, Size: 1l

New Product


Supplier: Honeywell Research Chemicals
Description: Chromasolv* plus Butyl acetate, Purity: 99.7%, Cas: 123-86-4, Molecular Formula: CH3COO(CH2)3CH3, Molar Mass: 116.16 g/mol, Synonym: High Purity Solvent, Liquid Chromatography, Container: Glass bottle, Color/Form: Colorl
Supplier: BeanTown Chemical
Description: N-Butyl acetate, Grade: Semiconductor, Purity: 99% minimum, Cas Number: 123-86-4, Molecular Formula: C6H12O2, Synonyms: Acetic acid n-butyl ester, Size: 4L

SDS

Catalog Number: (100503-736)
Supplier: Electron Microscopy Sciences
Description: Reagent, A.C.S. CH3COO(CH2)3CH3 F.W. 116.16 CAS #123-86-4 b.p . 127[degree]C Specific Gravity: 0.88

Minority or Woman-Owned Business Enterprise


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 18
no targeter for Bottom