    Sort by:
  • Search Within Results
Isopropyl acetate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"12"
  List View Easy View
Sort by:
Supplier:  Avantor Performance Materials
Description:   Lot analysis on label.
SDS
Supplier:  MilliporeSigma
Description:   Succinic Acid For Synthesis^
SDS
Catalog Number: (EM-PX1805-NP20)
Supplier:  MilliporeSigma
Description:   Isopropyl Acetate^
SDS
Supplier:  Alfa Aesar
Description:   Acetic acid isopropyl ester. Grade:99+, Melting Point C-73*. Boiling Point C:87-89*. C5H10O2. 108-21-4. HIGHLY FLAMMABLE IRRITANT
SDS
Supplier:  TCI America
Description:   CAS Number: 108-21-4 MDL Number: MFCD00008877 Molecular Formula: C5H10O2 Molecular Weight: 102.13 Purity/Analysis Method: 99.0% (GC) Form: Cl...
SDS
Product and/or packaging is safer to human and environmental health
Supplier:  Spectrum Chemicals
Description:   Acetic Acid Isopropyl Ester. Isopropyl Acetate. CAS: 108-21-4. Size: 200 Liter.
SDS
Small Business Enterprise
Supplier:  BeanTown Chemical
Description:   CAS: 108-21-4; EC No: 203-561-1; MDL No: MFCD00008877; RTECS: AI4930000 UN No: UN1220; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CO2C...
SDS
Supplier:  Honeywell Research Chemicals
Description:   Isopropyl acetate, Purity: >/=99.6%, Grade: Reagent, Cas number: 108-21-4, Molecular Formula: CH3COOCH(CH3)2, Molar mass: 102.13 g/mol, Density: 0.872 g/mL, Synonyms: IPrOAc; Acetic acid isopropyl ester, Performance Grade Solvent, General Purpose Reagent, Size: 1L
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 12
  1  2  Next 