    Sort by:
  • Search Within Results
Methyl ethyl ketone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"40"
Sort by:
Catalog Number: (BDH1137-19L)
Supplier:  VWR International
Description:   Meets ACS specifications for general use.
SDS
Catalog Number: (BDH25643.400)
Supplier:  VWR International
Description:   BDH* Methyl Ethyl Ketone, Chemical Formula: C4H8O, Melting Point: -86.6, Boiling Point: 79.5, Flash Point: -9 deg C, Formula Weight: 72.11, CAS Number: 78-93-3, Grade: HPLC/Spectro, HiPerSolv CHROMANORM, Class: F, Size: 4L
Supplier:  Avantor Performance Materials
Description:   2-Butanone. CAS RN 78-93-3. Formula Weight: 72.11. Density: 0.80kg/L. AR*, ACS, 99.0% min. by GC. Color (APHA): 15. Residue after Evaporation: 0.0025%. Titratable Acid: 0.0005 mEq/g. Water: 0.20%. Subject to postal/shipping restrictions. 1L.
SDS
Supplier:  Avantor Performance Materials
Description:   The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are es...
SDS
Catalog Number: (JT9214-3)
Supplier:  Avantor Performance Materials
Description:   Packaged under nitrogen. Lot analysis on label. For high performance liquid chromatography. UV cutoff 330nm.
SDS
Supplier:  Avantor Performance Materials
Description:   For use as an HPLC and TLC solvent. Filtered through a 0.2µm element.
SDS
Catalog Number: (JT9414-3)
Supplier:  Avantor Performance Materials
Description:   2-Butanone. CAS RN 78-93-3. Formula Weight: 72.11. Density: 0.80kg/L. CMOS* grade, 99.5% (by GC). Color (APHA): 10 max. PO4: 1ppm max. Residue after evaporation: 20ppm max. H2O (Karl Fischer): 0.2% max. Packaged in a glass bottle. Subject to postal/shipping restrictions. 3.79L.
SDS
Catalog Number: (CA71007-782)
Supplier:  AVANTOR PERF MAT - BDH
Description:   Methyl Ethyl Ketone Acs 4L^
SDS
Limited Availability
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 40
  1  2  3  4  5  Next