You Searched For: 4-Octanone
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (TCO0218-001ML)
Supplier: TCI America
Description: CAS Number: 589-63-9 MDL Number: MFCD00027241 Molecular Formula: C8H16O Molecular Weight: 128.22 Purity/Analysis Method: 98.0% (GC) Form: Cle...