    Sort by:
  • Search Within Results
4-dodecanone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   n-Octyl n-propyl ketone. Grade: 99, Melting Point C. Boiling Point C: 65*/0. 25mm. C12H24O. 6137-26-4.