Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "cryolite"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   MDL: MFCD00003507
Common Applications: Grain mounts, XRD and microprobe standards, identification of unknown minerals.
Notes: Sp. Gr. 2.90