    Sort by:
  • Search Within Results
hydrocortisone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"8"
Sort by:
Catalog Number: (MK883005)
Supplier:  Avantor Performance Materials
Description:   Assay (C21H30O5) (dried basis): 97. 0 - 102. 0 %. Chromatographic Purity: Passes Test. Identification A: Passes Test. Identification B: Passes Test. Loss on Drying: max. 1. 0 %. Organic Volatile Impurities: Passes Test. Residue on Ignition (mg): max.
Supplier:  Alfa Aesar
Description:   98%. 1g.
SDS
Supplier:  Spectrum Chemicals
Description:   BSE/TSE statement (available upon request) Allergen statement (available upon request)
Small Business Enterprise
Supplier:  MP Biomedicals
Description:   Soluble (mg/mL) at 25°C in: water 0.28; ethanol 15.0; methanol 6.2; acetone 9.3; chloroform 1.6; propylene glycol 12.7; ether ~ 0.35. Soluble in conce...
SDS
Catalog Number: (75810-948)
Supplier:  Spectrum Chemicals
Description:   HYDROCORTISONE, MICRONIZED, USP, otherwise known as Cortisol, is a glucocorticoid, a steroid hormone. Secreted in the adrenal cortex its function is t...
Supplier:  BeanTown Chemical
Description:   CAS: 50-23-7; EC No: 200-020-1; MDL No: MFCD00011654; RTECS: GM8925000 Crystalline; Molecular Formula: C21H30O5; MW: 362.47 Melting Point: 211-214°
SDS
Catalog Number: (101095-150)
Supplier:  US Pharmacopeia
Description:   (200 mg)
SDS
Supplier:  Acros Organics
Description:   Hydrocortisone, Purity: 98%, CAS No. 50-23-7, Molecular Formula: C21 H30 O5, MW: 362.47, Form: Powder Solid, Color: White, Synonym: 11beta,17alpha,21-Trihydroxypregn-4-ene-3,20-dione, Size: 5G
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8