    Sort by:
  • Search Within Results
2-Bromooctane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 557-35-7 MDL Number: MFCD00040823 Molecular Formula: C8H17Br Molecular Weight: 193.13 Purity/Analysis Method: 85.0% (GC) Form: Cl...