    Sort by:
  • Search Within Results
1-Bromoheptadecane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 3508-00-7 MDL Number: MFCD00017891 Molecular Formula: C17H35Br Molecular Weight: 319.37 Purity/Analysis Method: 98.0% (GC) Form: ...