    Sort by:
  • Search Within Results
1-Bromodocosane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 6938-66-5 MDL Number: MFCD00013543 Molecular Formula: C22H45Br Molecular Weight: 389.51 Purity/Analysis Method: 98.0% (GC) Form: ...