    Sort by:
  • Search Within Results
Sodium selenate decahydrate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   99.9% (metals basis),UN2630,Crystalline,WARNING: Causes garlic odor on breath; eye & lung irritation,25g,100g,POISON,DOT-E8249