tetra-Sodium diphosphate decahydrate
Supplier:
Avantor Performance Materials
Description:
Crystals. Lot analysis on label.
Supplier:
MilliporeSigma
Description:
EMSURE* tetra-Sodium diphosphate decahydrate for analysis 2.5kg^
Supplier:
Alfa Aesar
Description:
ACS, 99.0-103.0%.Crystalline,WARNING: Causes GI injury, skin and eye irritation,500g,100g.
Supplier:
BeanTown Chemical
Description:
CAS: 13472-36-1; EC No: 231-767-1; MDL No: MFCD00149200; RTECS: UX7350000
Crystalline; Linear Formula: Na4P2O7·10H2O; MW: 446.06
Melting Point: 79.5°
...
Supplier:
Acros Organics
Description:
Sodium pyrophosphate decahydrate, Purity: 99-13%, CAS Number: 13472-36-1, Molecular Formula: Na4O7P2.10H2O, Formula Weight: 446.05g/mol, Synonyms: TSPP, Physical Form: Crystals, Color: White, Size: 2.5kg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
* Please note if your account is within the State of California two of these pieces of identification will be required.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
|
|||||||||