You Searched For: Sodium carbonate monohydrate


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Sodium Carbonate, Monohydrate, Granular, AR® (ACS)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Sodium Carbonate, Monohydrate, Crystal, N.F., Multi-Compendial
Catalog Number: (VWRB427-12KG)
Supplier: Avantor Performance Materials
Description: Tested according to the NF, EP, and BP monographs. Material is of non-animal, non-human origin.

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Sodium Carbonate, Monohydrate, Crystal, BAKER ANALYZED® A.C.S. Reagent
Supplier: Thermo Scientific Chemicals
Description: 85% min.Crystalline,WARNING: Irritates skin and eyes,1kg,5kg.
Supplier: Thermo Scientific Chemicals
Description: ACS, 99.5% min.Granular,WARNING: Irritates skin and eyes,500g,2kg.
Supplier: Spectrum Chemicals
Description: Sodium Carbonate, Hydrous, NF is a pH regulator excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current...
Supplier: Spectrum Chemicals
Description: Sodium Carbonate, Monohydrate, FCC is a food additive (E500) used as an acidity regulator, anti-caking agent, raising agent, and stabilizer. The FCC g...
Catalog Number: (BJ230952-500G)
Supplier: Honeywell Research Chemicals
Description: Sodium carbonate monohydrate, ACS Reagent, CAS Number: 5968-11-6, Purity: >/= 99.5%, Linear Formula: Na2CO3 a.H2O, Molar Mass: 124.00 g/mol, Laboratory Chemical, ACS Grade Salt, Size: 500G

SDS


Supplier: BeanTown Chemical
Description: Sodium carbonate monohydrate, ACS, Purity: 99.5% min, Cas number: 5968-11-6, Molecular Formula: CH2Na2O4, Size: 2KG

SDS

Supplier: Thermo Scientific Chemicals
Description: SODIUM CARBONATE MONOHYDRATE 5KG

Supplier: MilliporeSigma
Description: 25kg
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 13 of 13
no targeter for Bottom