    Sort by:
  • Search Within Results
You Searched for: "r-1-aminoindan"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   (R)-(-)-1-Indanamine. Grade: ChiPros\(tm 99+%, ee 98+. Melting Point C0-2*. Boiling Point C: 96-98*/8mm. C9H11N. 10277-74-4. Trademark of BASF AG. IRRITANT AIR SENSITIVE