    Sort by:
DL-Camphor
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"8"
  List View Easy View
Sort by:
Supplier:  MilliporeSigma
Description:   DL-CAMPHOR FOR SYNTHESI (250G).
SDS
Supplier:  Alfa Aesar
Description:   97+%. 500g.
SDS
Supplier:  TCI America
Description:   CAS Number: 76-22-2 MDL Number: MFCD00074738 Molecular Formula: C10H16O Molecular Weight: 152.24 Purity/Analysis Method: 95.0% (GC) Form: Cry...
SDS
Supplier:  Spectrum Chemicals
Description:   BSE/TSE statement (available upon request) Allergen statement (available upon request)
SDS
Small Business Enterprise
Catalog Number: (700003-996)
Supplier:  Spectrum Chemicals
Description:   DL-Camphor. CAS: 76-22-2. Size: 1 LB.
SDS
Supplier:  Spectrum Chemicals
Description:   CAMPHOR, SYNTHETIC, USP is a white to transparent solid that has a waxy texture, strong aromatic odor and flammable. It is found naturally in the wood...
Supplier:  BeanTown Chemical
Description:   CAS: 76-22-2; EC No: 200-945-0; MDL No: MFCD00074738; RTECS: EX1225000 UN No: UN2717; Haz Class: 4.1; Packing Group: III Solid; Molecular Formula: C10...
SDS
Supplier:  Acros Organics
Description:   DL-Camphor, Purity: 96%, CAS No 76-22-2, Molecular weight: 152.24, Molecular formula: C10 H16 O, Appearance (, Color): White, Appearance (Form): Crystals and/or chunks, Size: 250G
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8