Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "spades-and-shovels"

https://us.vwr.com/stibo/smallweb/std.lang.all/98/19/18989819.jpg

High-quality spades and shovels efficiently move materials in various volume sizes. Cost efficient alternatives to electric shovels when dealing with small quantities, the tools have resistant manufacturing and unbreakable thickness to handle any load. Featuring ergonomic handhold grips that assist with lifting, the one-piece diggers are available in color coded and wide blade options. Adaptable to any task or environment, the spades and shovels can be used at home or meet food industry regulations.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"22"
Sort by:
 
 
 
 

Supplier:  Remco Products
Description:   Remco one-piece polypropylene shovels are tough, lightweight and hygienic. Molded from FDA-compliant polypropylene, it is a workhorse for moving bulk ...
Catalog Number: (470151-768)
Supplier:  ARETT SALES CORP
Description:   These Garden Trowels are perfect for plant experiments and activities.
Catalog Number: (470145-126)
Supplier:  HB DAVIS SEED CO. INC
Description:   Versatile Tool.
Catalog Number: (470013-336)
Supplier:  LIBERTY MOUNTAIN SPORTS, LLC
Description:   This compact and lightweight folding shovel can be used at a 90° angle for chopping or straight for shovelling, or it can be securely locked in three ...
Supplier:  Remco Products
Description:   Remco one-piece polypropylene shovels are tough, lightweight and hygienic. Molded from FDA-compliant polypropylene, it is a workhorse for moving bulk ...
Supplier:  Remco Products
Description:   Remco one-piece polypropylene shovels are tough, lightweight and hygienic. Molded from FDA-compliant polypropylene, it is a workhorse for moving bulk ...
Supplier:  Remco Products
Description:   This compact shovel is ideal for spill kit applications, as well as a wide variety of other uses. Constructed of injection-molded ABS plastic, it will...
Supplier:  Remco Products
Description:   This Remco one-piece polypropylene shovel is tough, lightweight and ergonomic. It is molded from FDA-compliant materials, and features a second grip t...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 22
  1  2  3  Next