Dataproducts® R73402 Typewriter Ribbon

DPSR73402
500023-971EA 13.76 USD
500023-971
Dataproducts® R73402 Typewriter Ribbon
For use with typewriter.

  • Long life and exceptional character definition
  • Typewriter device type
  • Correctable ribbon type
  • Compatible for use

Color: Black.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR