Would you like to log in again?
Swerve Banded Hearing Protector
Supplier: Janitorial Supplies
E·A·R®
MMM3222000
Banded hearing protector features a flexible, comfortable band which helps keep ear plugs in place
Includes two sets of ear plug tips: Flex 28 tips and Comfort Pod tips that do not penetrate the ear canal as far with an NRR of 19 dB.
Ordering information: The 3M products in this catalog must be used in accordance with OSHA regulations, user instructions, warnings, and limitations that accompany each item.
ORDER
Expand All / Collapse All
Color Cord Noise Reduction Rate (NRR) Supplier No. VWR Catalog Number Unit Price Quantity
Yellow Corded 28 dB MMM3222000 500022-593 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty